Sino Biological™ IL18R1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 329) of human IL18R1 (NP_003846.1) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11102H08H5

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13496106

Quantity Price
1 £ 54.5 / 5µg
Estimated Shipment date
from Supplier 18-04-2017
Add to basket


Description and Specification

Specification

Gene Alias IL18R1, CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
Molecular Weight 37.2kDa
Format Lyophilized
Gene ID (Entrez) 8809
Expression System Mammalian
Protein Family Cytokines & Receptors
Protein Subtype Interleukins|Cytokine Receptors
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Quantity 5μg
Protein Tag His Tag
Structural Form Truncated/Recombinant
Common Name Interleukin 18 receptor 1
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol IL18R1

Interleukin 18 Receptor 1 (IL18R1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.