missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL18R1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 329) of human IL18R1 (NP_003846.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11102H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486106

  • £425.77 / 250µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Interleukin 18 Receptor 1 (IL18R1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8809
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL18R1, CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
IL18R1
Cytokines & Receptors
His Tag
Lyophilized
37.2kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 18 receptor 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.