missing translation for 'onlineSavingsMsg'
Learn More

IL18BP Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the precursor (Met 1-Ala 193) of mouse IL18BP (NP_034661.1) fused to a polyhistide tag at the C-terminus

Brand:  Invitrogen 50206M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13446256

  • £494.00 / 250µg

Estimated Shipment date
from Supplier 31-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 18 binding protein (IL18BP), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse IL18BP-His at 1μg/well can bind biotinylated cynomolgus IL18 with a linear range of 0.16-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16068
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins
Il18bp, IL-18BP, Igifbp, MC54L
IL8BP
Cytokines & Receptors
His Tag
Lyophilized
19.4kDa
5 x 50μg
Recombinant/Ligand Binding Domain
<1 EU/μg
Interleukin 18 binding protein
Assay Standard
Mammalian
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.