missing translation for 'onlineSavingsMsg'
Learn More

IL18BP Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the N-terminal domain (Met 1-Gly 192) of human IL18BP (NP_001034748.1) fused to the polyhistidine tag at the C-terminus

Brand:  Invitrogen 10357H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435836

  • £222.00 / 50µg

Estimated Shipment
03-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 18 binding protein (IL18BP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL18BP Protein (His Tag) at 1μg/well can bind biotinylated cynomolgus IL18 with a linear range of 0.16-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10068
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins
IL18BP, IL18BPa
IL8BP
Cytokines & Receptors
His Tag
Lyophilized
19kDa
50μg
Recombinant/Ligand Binding Domain
<1 EU/μg
Interleukin 18 binding protein
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.