missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL17RC Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the full length of human IL17RC isoform 3 (NP_116121.2), residues 1 to 454, fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11747H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405596

  • £210.29 / 50µg


Estimated Shipment
17-05-2018
Add to basket

Description

Description

Interleukin 17 Receptor C (IL17RC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris buffer, pH 7.4, 500mM NaCl, 10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
84818
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
UNQ6118, PRO20040, PRO38901, FLJ95963, FLJ96005, IL17-RL, IL17RL, mgC10763
IL17RC
Cytokines & Receptors
His Tag
Lyophilized
49.6kDa
50μg
Recombinant
<1 EU/μg
Interleukin 17 receptor C
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.