missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL16 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL16 (AAC12732.1) (Pro 2-Ser 130) fused to a polyhistide tag at the N-terminus

Brand:  Thermo Scientific™ 10944H07E5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15247435

  • £102.81 / 5µg

Estimated Shipment
Add to basket



Interleukin 16 (IL16), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human CD4-Fc in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Interleukin 16
Assay Standard
E. coli
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FLJ16806, FLJ42735, FLJ44234, LCF, NIL16, PRIL16, prIL-16
Cytokines & Receptors
His Tag

For Research Use Only. Not for use in diagnostic procedures.