missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL13RA2 Recombinant Mouse Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 334) of mouse IL13RA2 (NP_032382.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50061M03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13406186

  • £186.34 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Interleukin 13 receptor, alpha 2 (IL13RA2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL13 at 20μg/mL (100μL/well) can bind mouse IL13RA2 with a linear range of 0.5-2μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16165
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
Il13ra2, CD213a2
IL13RA2
Cytokines & Receptors
His Tag/IgG1 Fc Tag
Lyophilized
64.5kDa
50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 13 receptor, alpha 2
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.