missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL13RA1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human IL13RA1 (NP_001551.1) extracellular domain (Met 1-Thr 343) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10943H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15207435

  • £65.07 / 5µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Interleukin 13 Receptor Alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL13RA1 at 4μg/mL (100μL/well) can bind human IL13 with a linear ranger of 0.156-2.5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38.3kDa
5μg
Interleukins|Cytokine Receptors
IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4
IL13RA1
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 13 receptor alpha 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.