missing translation for 'onlineSavingsMsg'
Learn More

IL13RA1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 343) of human IL13RA1 (NP_001551.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

£57.75 - £496.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4
Gene ID (Entrez) 3597
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13476016 View Documents Invitrogen
10943H03H5
5μg £57.75
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
13446016 View Documents Invitrogen
10943H03H25
5 x 5μg £149.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
13456016 View Documents Invitrogen
10943H03H250
5 x 50μg £496.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interleukin 13 receptor, alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL13 at 4μg/mL (100μL/well) can bind human IL13RA1-Fc with a linear range of 0.156-2.5μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
3597
<1 EU/μg
IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4

For Research Use Only. Not for use in diagnostic procedures.