Learn More
IL13RA1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 343) of human IL13RA1 (NP_001551.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 10943H03H5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Interleukin 13 receptor, alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.- Activity: Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL13 at 4μg/mL (100μL/well) can bind human IL13RA1-Fc with a linear range of 0.156-2.5μg/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
65kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Interleukin 13 receptor, alpha 1 | |
His Tag/IgG1 Fc Tag | |
Cytokines & Receptors/CD Proteins | |
Interleukins, Cytokine Receptors |
3597 | |
5μg | |
Truncated/Recombinant | |
IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4 | |
IL13RA1 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.