missing translation for 'onlineSavingsMsg'
Learn More

IL13RA1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 343) of human IL13RA1 (NP_001551.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10943H03H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13456016

  • £496.00 / 250µg

Estimated Shipment
21-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 13 receptor, alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL13 at 4μg/mL (100μL/well) can bind human IL13RA1-Fc with a linear range of 0.156-2.5μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Interleukins|Cytokine Receptors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3597
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 13 receptor, alpha 1
His Tag/IgG1 Fc Tag
Cytokines & Receptors/CD Proteins
Assay Standard
Lyophilized
65kDa
5 x 50μg
Truncated/Recombinant
IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4
IL13RA1
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.