missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL13 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding human IL-13 (AAK53823.1) (Met 1 - Asn 132)

Brand:  Thermo Scientific™ 10369HNAC250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15285275

  • £1546.93 / 250µg


Estimated Shipment
15-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interleukin 13 (IL13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the ED50 for this effect is typically 0.75-3.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12.5kDa
5 x 50μg
Interleukins
ALRH, BHR1, IL-13, mgC116786, mgC116788, mgC116789, P600
IL13
Cytokines & Receptors
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 13
Assay Standard
Mammalian
Videos