Sino Biological™ IL12RB2 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse IL12RB2 (NP_032380.1) extracellular domain (Met 1-Asn 637) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 50099M08H50

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of protein in suspension buffer

Product Code. 15382716

Quantity Price
1 £ 197.0 / 50µg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Gene ID (Entrez) 16162
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Purity ≥95% by SDS-PAGE
Protein Family Cytokines & Receptors
Expression System Mammalian
Purification Method PAGE
Validated Application Assay Standard
Gene Alias A930027I18Rik, IL-12RB2, Ifnm
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 50μg
Endotoxin Concentration <1 EU/μg
Termination C-Terminal
Protein Tag His Tag
Molecular Weight 70kDa
Common Name Interleukin 12 receptor beta 2
Gene Symbol IL12RB2
Format Lyophilized
Structural Form Truncated/Recombinant

Interleukin 12 Receptor Beta 2 (IL12RB2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.