missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL12B Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the p40 subunit of human IL12, IL12B (NP_002178.2) (Met 1-Ser 328) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10052H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15264555

  • £373.72 / 25µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

Interleaukin 12 subunit beta (IL12B), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in a functional ELISA: 1. Measured by its ability to bind biotinylated recombinant human IL12RB1; 2. Measured by its ability to bind biotinylated Human IL12A-His
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.2kDa
5 x 5μg
Interleukins
IL12B, CLMF, CLMF2, IL-12B, NKSF, NKSF2
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
IL12B
Cytokines & Receptors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.