Learn More
IL10RB Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 220) of human IL10RB (NP_000619.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 10945H03H5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Interleukin 10 receptor, beta (IL10RB), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human IL28B at 20μg/mL (100μL/well) can bind human IL10RB-Fc with a linear range of 3.125-25μg/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
51.7kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Interleukin 10 receptor, beta | |
His Tag/IgG1 Fc Tag | |
Cytokines & Receptors | |
Interleukins, Cytokine Receptors |
3588 | |
5μg | |
Truncated/Recombinant | |
IL10RB, CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2 | |
IL10RB | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.