missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL10RB Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 220) of human IL10RB (NP_000619.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10945H03H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13496016

  • £620.44 / 250µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Interleukin 10 receptor, beta (IL10RB), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL28B at 20μg/mL (100μL/well) can bind human IL10RB-Fc with a linear range of 3.125-25μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3588
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL10RB, CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2
IL10RB
Cytokines & Receptors
His Tag/IgG1 Fc Tag
Lyophilized
51.7kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 10 receptor, beta
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.