Sino Biological™ IGFBP6 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP6 (P24592) (Met 1-Gly 240) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 13026H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15266105

Quantity Price
1 £ 80.25 / 5µg
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Specification

Protein Tag His Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 23.9kDa
Format Lyophilized
Protein Family Growth Factors & Receptors
Expression System Mammalian
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
Quantity 5μg
Endotoxin Concentration <1 EU/μg
Gene Symbol IGFBP6
Purity ≥90% by SDS-PAGE
Gene Alias IBP6
Common Name Insulin-like growth factor binding protein 6

Insulin-like Growth Factor Binding Protein 6 (IGFBP6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human IGF1 in functional ELISA; 2. Measured by its ability to bind human IGF2 in functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.