missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGFBP4 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP4 (NP_001543.2) precursor (Met 1-Glu 258) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50250M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15313006

  • £1016.02 / 250µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Description

Description

Insulin-Like Growth Factor Binding Protein 4 (IGFBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a serum-free cell proliferation assay using MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. 1988, Cancer Research 48: 4083); the ED50 is typically 5-10μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27.2kDa
5 x 50μg
IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
Igfbp4, RP23-333D2.6, AI875747, Deb2, IGFBP-4
IGFBP4
Growth Factors & Receptors
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Insulin-like growth factor binding protein 4
Assay Standard
Mammalian
Videos