missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGFBP4 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP4 (NP_001543.2) precursor (Met 1-Glu 258) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10967H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15287455

  • £200.92 / 25µg


Estimated Shipment
14-05-2018
Add to basket

Description

Description

Insulin-Like Growth Factor Binding Protein 4 (IGFBP4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human IGF1 (Mfr. No. 10598-H24E) in functional ELISA; 2.Measured by its ability to bind human IGF2 (Mfr. No. 13032-H24E) in functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27.4kDa
5 x 5μg
IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
BP-4, HT29-IGFBP, IBP4, IGFBP-4, IGFBP4
IGFBP4
Growth Factors & Receptors
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Insulin-like growth factor binding protein 4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.