missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGFBP3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGFBP3 (NP_000589.2), residues 28 to 291, fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10430H07H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465876

  • £68.72 / 5µg


Estimated Shipment
05-06-2018
Add to basket

Description

Description

Insulin-like growth factor binding protein 3 (IGFBP3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human IGF2 at 10μg/mL (100μL/well) can biotinylated human IGFBP3-His, the ED50 of can biotinylated human IGFBP3-His is 18ng/mL; 2. Immobilized human IGF1 at 10μg/mL (100μL/well) can biotinylated human IGFBP3-His, the ED50 of can biotinylated human IGFBP3-His is 24ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3486
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IGFs (Insulin-Like Growth Factors)
IGFBP3, tcag7.703, BP-53, IBP3
IGFBP3
Growth Factors & Receptors
His Tag
Lyophilized
31kDa
5μg
Recombinant
<1 EU/μg
Insulin-like growth factor binding protein 3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.