missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGF2BP2 Recombinant Human Protein, N-His.GST Tag

This recombinant protein was expressed from a DNA sequence encoding the human IGF2BP2 isoform 1 (Q9Y6M1-2) (Met 1-Lys 599) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Brand:  Thermo Scientific™ 11116H20B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228255

  • £80.42 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Insulin-like Growth Factor 2 mRNA Binding Protein 2 (IGF2BP2), recombinant human protein is supplied as a lyophilized powder.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 500mM NaCl , 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10644
PAGE
5μg
N-Terminal
<1 EU/μg
Insulin-like Growth Factor 2 mRNA Binding Protein 2
Assay Standard
Insect
Lyophilized
94kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
IMP-2, IMP2, VICKZ2, p62
IGF2BP2
Growth Factors & Receptors
GST Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.