missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGF2 Recombinant Human Protein, His.NusA Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF2 (P01344-1) (Ala 25-Glu 91) fused to the polyhistidine-NusA tag at the N-terminus

Brand:  Thermo Scientific™ 13032H24E5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13413079

  • £55.70 / 5µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Insulin-like growth factor 2 (IGF2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF2 at 10μg/mL (100μL/well) can bind biotinylated human His-IGFBP3; the EC50 of binding to biotinylated human His-IGFBP3 is 18ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
64.6kDa
5μg
Full Length/Recombinant
PP1446, C11orf43, FLJ22066, FLJ44734, IGF-II, PP9974
IGF2
Growth Factors & Receptors
His Tag, NusA Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IGFs (Insulin-Like Growth Factors)
Insulin-like growth factor 2 (somatomedin A)
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.