missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IGF1 Recombinant Human Protein, His.NusA Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF1 isoform 1 (P05019-1) (Gly 49-Ala 118) fused to the polyhistidine-NusA tag at the N-terminus

Brand:  Thermo Scientific™ 10598H24E25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15266975

  • £141.58 / 25µg

Estimated Shipment
Add to basket



Insulin-like growth factor 1 (IGF1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF1 at 10μg/mL (100μL/well) can bind biotinylated human IGFBP3-His, the EC50 of biotinylated human IGFBP3-His is 24ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
Full Length/Recombinant
Growth Factors & Receptors
His Tag, NusA Tag
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IGFs (Insulin-Like Growth Factors)
Insulin-like growth factor 1 (somatomedin C)
Assay Standard
E. coli

For Research Use Only. Not for use in diagnostic procedures.