Sino Biological™ IFNGR1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 245) of human IFN-γ R1 pre-protein (NP_000407.1) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10338H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15265215

Quantity Price
1 £ 54.75 / 5µg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Purity ≥90% by SDS-PAGE
Gene Alias IFNGR1, RP3-503F13.3, CD119, FLJ45734, IFNGR
Structural Form Truncated/Recombinant
Purification Method PAGE
Common Name Interferon gamma receptor 1
Validated Application Assay Standard
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 3459
Format Lyophilized
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Protein Family Cytokines & Receptors/CD Proteins
Molecular Weight 27.3kDa
Termination C-Terminal
Gene Symbol IFNGR1
Protein Tag His Tag
Quantity 5μg

Interferon Gamma Receptor 1 (IFNGR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.