missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IFNG Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse IFNG (P01580) (Met 1-Cys 155) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50709M02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15389196

  • £82.76 / 25µg


Estimated Shipment
15-06-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Interferon gamma (IFNG) recombinant mouse protein is supplied as a lyophilized powder
  • Activity: Measured in anti-viral assays using L929 cells infected with vesicular stomatitisvirus (VSV); the ED50 for this effect is typically 20–80ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween® 80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
15978
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Interferon gamma
Assay Standard
Mammalian
Lyophilized
42.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
IFN-g, Ifg
IFNG
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.