missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IFNB1 (INF-β) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human IFNB1 (NP_002167.1), residues 1 to 187, fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10704H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13495946

  • £239.43 / 50µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interferon, beta 1 (IFNB1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the ED50 for this effect is 0.5-2ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3456
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interferons
IFNB1, IFB, IFF, IFNB, mgC96956
IFNB1
Cytokines & Receptors
IgG1 Fc Tag
Lyophilized
46.7kDa
50μg
Recombinant
<1 EU/μg
Interferon, beta 1, Fibroblast
Assay Standard
Mammalian
Videos