missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IFNAR2 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the human IFNAR2 isoform a (NP_997468.1) (Met 1-Lys 243) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10359H02H5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13482979

  • £56.74 / 5µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Interferon (alpha, beta and omega) receptor 2 (IFNAR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit rhesus IFNB-mediated protection of WISH human amnion cells infected with vesicular stomatitis virus (VSV) to viral lysis, the EC50 for this effect is typically 0.08-0.4μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
51.8kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Interferon (alpha, beta and omega) receptor 2
Assay Standard
Mammalian
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interferons
IFN-R, IFN-alpha-REC, IFNABR, IFNARB
IFNAR2
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.