missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IFNAR1 Recombinant Human Protein, IgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human IFNAR1 isoform 1 (P17181-1) extracellular domain (Met 1- Lys 436) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 13222H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15266265

  • £172.81 / 25µg


EU Stock 1
Add to basket

Description

Description

Interferon (alpha, beta and omega) Receptor 1 (IFNAR1), recombinant human protein is supplied as a lyophilized powder.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3454
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Interferon (alpha, beta and omega) Receptor 1
Assay Standard
Mammalian
Lyophilized
74kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
IFNAR1
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.