missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IFNA4 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IFNA4 (NP_066546.1) (Cys 23-Asp 189) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10336H01H5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13432969

  • £56.74 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Interferon, alpha 4 (IFNA4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the EC50 for this effect is 0.8-4ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
46kDa
5μg
Full Length/Recombinant
<1 EU/μg
Interferon, alpha 4
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interferons
INFA4, mgC142200
IFNA4
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.