Learn More
ICOSLG Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mouse ICOSL (NP_056605.1) extracellular domain (Met 1-Lys 279) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 50190M03H50
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: One vial of lyophilized protein
Description
Inducible T-Cell Co-Stimulator Ligand (ICOSL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human ICOS at 1μg/mL (100μL/well) can bind mouse ICOSL with a linear range of 0.125-1.0μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50μg | |
<1 EU/μg | |
Inducible T-cell co-stimulator ligand | |
His Tag/IgG1 Fc Tag | |
CD Proteins | |
Other Proteins |
54.3kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Icosl, AU044799, B7-H2, B7RP-1, B7h, BG071784, GI50, GL50, GL50-B, ICOS-L, Icoslg | |
ICOSL | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.