missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ICOSLG (ICOSL) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human ICOSLG (NP_056074.1) extracellular domain (Met 1-Ser 258) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11559H08H5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15208665

  • £57.26 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Inducible T-cell co-stimulator ligand (ICOSLG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human ICOSLG at 1μg/mL (100μL/well) can bind human ICOS with a linear range of 1.6-200ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28kDa
5μg
Other Proteins
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS
ICOSLG (ICOSL)
CD Proteins
His Tag
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Inducible T-cell co-stimulator ligand
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.