missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human ICAM1 (NP_000192.2) (Met 1 - Glu 480) fused to a polyhistidine tag at the C-terminus
£147.00 - £189.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 51kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Truncated/Recombinant/Cell Surface Protein |
Endotoxin Concentration | <1 EU/μg |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13402979
|
Invitrogen
10346H08H25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13432979
|
Invitrogen
10346H08H50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Intercellular adhesion molecule 1 (ICAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured by the ability of the immobilized protein to support the adhesion of PMA stimulated HSB2 human peripheral blood acute lymphoblastic leukemia cells; when cells are added to ICAM1 coated plates (12.5μg/mL, 100μL/well), approximately 20-40% will adhere specifically
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Intercellular adhesion molecule 1 | |
His Tag | |
CD Proteins | |
Intercellular Adhesion Molecule |
51kDa | |
Truncated/Recombinant/Cell Surface Protein | |
BB2, CD54, P3.58 | |
ICAM1 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.