missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ICAM1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human ICAM1 (NP_000192.2) (Met 1 - Glu 480) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10346H08H5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13412979

  • £56.47 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Intercellular adhesion molecule 1 (ICAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of PMA stimulated HSB2 human peripheral blood acute lymphoblastic leukemia cells; when cells are added to ICAM1 coated plates (12.5μg/mL, 100μL/well), approximately 20-40% will adhere specifically
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
51kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Intercellular adhesion molecule 1
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Intercellular Adhesion Molecule
BB2, CD54, P3.58
ICAM1
CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.