missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HTRA2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human HTRA2 (O43464-1) (Ala 134-Glu 458) fused to a polyhistide tag at the C-terminus

Brand:  Thermo Scientific™ 10619H08E25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15216995

  • £127.01 / 25µg


Estimated Shipment
14-05-2018
Add to basket

Description

Description

HTRA Serine Peptidase 2 (HTRA2), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Protease activity demonstrated by HTRA2 cleavage of bovine beta-casein (Sigma Cat. No. C-6905); incubation of beta-casein at 0.2μg/mL with Recombinant Human HTRA2 at 0.02μg/mL (ratio of 10:1) for 60 minutes at 45°C in 50mM Tris, pH 8.0, which results in >95% cleavage of beta-casein, as revealed by SDS-PAGE
  • Formulation: 50mM Tris, 0.3M NaCl, 1mM DTT, 0.2μg/μL BSA, 20% glycerol, pH 7.8
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.5kDa
5 x 5μg
Proteases
HtrA serine peptidase 2
Assay Standard
His Tag
Frozen
≥85% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
OMI, PARK13, PRSS25
HTRA2
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.