missing translation for 'onlineSavingsMsg'
Learn More

HPX Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mouse HPX (NP_059067.2) (Met 1 - Gln 460) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 50271M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15343036

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: One vial of lyophilized protein

Description

Description

Hemopexin (HPX), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind protoporphyrin IX (PPPIX); Recombinant mouse Hemopexin (HPX) binds >15μM PPPIX, resulting in a 50% decrease in the fluorescence signal of mouse HPX
  • Formulation: Lyophilized in 20mM MES, 150mM NaCl, pH 6.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Hemopexin
His Tag
Other Proteins
Other Proteins
50.4kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Hpxn, hx, HPX
HPX
Mammalian
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.