missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HPX Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse HPX (NP_059067.2) (Met 1 - Gln 460) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50271M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15323036

  • £142.62 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Hemopexin (HPX), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind protoporphyrin IX (PPPIX); Recombinant mouse Hemopexin (HPX) binds >15μM PPPIX, resulting in a 50% decrease in the fluorescence signal of mouse HPX
  • Formulation: Lyophilized in 20mM MES, 150mM NaCl, pH 6.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50.4kDa
5 x 5μg
Other Proteins
Hpxn, hx, HPX
HPX
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Hemopexin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.