missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HPGD Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human HPGD (NP_000851.2) (Met 1-Gln 266) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11205H08E5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15228375

  • £142.62 / 5µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Hydroxyprostaglandin Dehydrogenase 15-(Nad) (HPGD), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measure by the production of NADH during the oxidation of PGF2-alpha; the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM DTT, 0.2μg/μL BSA, 10% glycerol, pH.7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
29.7kDa
5μg
Other Proteins
Hydroxyprostaglandin dehydrogenase 15-(NAD)
Assay Standard
His Tag
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
15-PGDH, PGDH, PGDH1, SDR36C1, HPGD
HPGD
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.