Sino Biological™ HIST3H2A Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human HIST3H2A (NP_254280.1) (Met 1-Lys 130)

Overview
Brand: Invitrogen

Manufacturer Part Number: 10926HNAE5

HIST3H2A RECOMBINANT HUMAN PROTEIN

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15277395

Quantity Price
1 £ 80.5 / 5µg
EU Stock 3
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration <1 EU/μg
Protein Family Histones
Gene ID (Entrez) 92815
Molecular Weight 14.2kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5μg
Structural Form Full Length/Recombinant
Protein Subtype Histones
Gene Alias MGC3165
Common Name Histone cluster 3, H2a
Gene Symbol HIST3H2A
Validated Application Assay Standard
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Histone Cluster 3, H2A (HIST3H2A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM β-Mercaptoethanol, pH 6.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.