missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HIST2H2BE Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human HIST2H2BE (Q16778) (Met 1-Lys 126)

Brand:  Thermo Scientific™ 11362HNAE50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238555

  • £539.24 / 50µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Histone Cluster 2, H2BE (HIST2H2BE), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, pH 6.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8349
PAGE
50μg
Full Length/Recombinant
GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, mgC119802, mgC119804, mgC129733
HIST2H2BE
Histones
Lyophilized
14.2kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Histone Cluster 2, H2BE
Assay Standard
E. coli
Videos