Sino Biological™ HIST2H2BE Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human HIST2H2BE (Q16778) (Met 1-Lys 126)

Overview
Brand: Invitrogen

Manufacturer Part Number: 11362HNAE25

HIST2H2BE RECOMBINANT HUMAN PROTEIN

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15208555

Quantity Price
1 £ 329.0 / 25µg
Estimated Shipment date
from Supplier 08-03-2017
Add to basket


Description and Specification

Specification

Validated Application Assay Standard
Protein Family Histones
Molecular Weight 14.2kDa
Purification Method PAGE
Purity ≥95% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Protein Subtype Histones
Endotoxin Concentration <1 EU/μg
Gene Alias GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, mgC119802, mgC119804, mgC129733
Common Name Histone Cluster 2, H2BE
Gene Symbol HIST2H2BE
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 8349

Histone Cluster 2, H2BE (HIST2H2BE), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, pH 6.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.