missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1, (A/Vietnam/1203/2004), mIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of influenza A virus, subtype H5N1 (A/VietNam/1203/2004), HA1 subunit (AAW80717.1) fused to the His-tagged Fc region of mouse IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10003V06H125

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445656

  • £141.31 / 25µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

H5N1, recombinant influenza A virus protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
26.1kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian
Lyophilized
≥85% by SDS-PAGE and SEC-HPLC Analysis
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA1
HA
Other Proteins
His Tag/IgG1 Fc Tag
Videos