missing translation for 'onlineSavingsMsg'
Learn More

HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Anhui/1/2005), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Pro 337) of influenza A virus, subtype H5N1 (A/Anhui/1/2005), HA1 subunit (ABD28180.1) fused to a polyhistidine tag at the C-terminus

£75.75 - £680.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13446046 View Documents Invitrogen
11048V08H25
5μg £75.75
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-04-2020
Please to purchase this item. Need a web account? Register with us today!
13426046 View Documents Invitrogen
11048V08H225
5 x 5μg £144.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-04-2020
Please to purchase this item. Need a web account? Register with us today!
13456046 View Documents Invitrogen
11048V08H250
50μg £221.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-04-2020
Please to purchase this item. Need a web account? Register with us today!
13436046 View Documents Invitrogen
11048V08H2250
5 x 50μg £680.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
HA1

For Research Use Only. Not for use in diagnostic procedures.