missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H3N2 (A/Aichi/2/1968), His Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 345) of the influenza A virus, subtype H3N2 (A/Aichi/2/1968), HA1 (AAA43178.1) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 11707V08H1250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465646

  • £662.89 / 250µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Hemagglutinin, Influenza A Virus, HA1 Subunit (HA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37.6kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA1
HA
Other Proteins
His Tag
Videos