missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H1N1 (A/California/04/2009), His Tag

This recombinant protein was expressed from a DNA sequence encoding an N-terminal segment (Met 1-Arg 344) of influenza A virus, subtype H1N1 (A/California/04/2009), HA1 (ACP41105.1) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 11055V08H45

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13466066

  • £74.55 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Hemagglutinin, Influenza A Virus (HA1 Subunit), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA1
HA
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.