missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA1 Recombinant Influenza A Virus Protein Subtype H1N1 (A/England/195/2009), His Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 344) of influenza A H1N1 (A/England/195/2009 (H1N1)) hemagglutinin (ACR15621.1) termed as HA1, and this protein has a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 40005V08H125

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15296375

  • £141.31 / 25µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

Hemagglutinin A, Subunit 1 (HA1), recombinant Influenza A H1N1 (A/England/195/2009) HA1 protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Unknown
PAGE
5 x 5μg
C-Terminal
Viral Proteins|Glycoproteins
HA1
HA1
Cell Structure/Organelle Proteins
His Tag
Lyophilized
37.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
<1 EU/μg
Hemagglutinin A, subunit 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.