missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H7N7 (A/Netherlands/219/03), His Tag

This recombinant protein was expressed from a DNA sequence encoding the native extracellular domain (Met 8-Phe 544) of influenza A virus, subtype H7N7 (A/Netherlands/219/03), HA (AAR02640.1) (Met 8-Phe 544), fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11082V08B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13406096

  • £141.31 / 25µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in hemagglutination test using 1% chicken erythrocyte; the HA titer is 0.1-0.5μg/mL for 1% chicken red blood cells
  • Formulation: Lyophilized in 20mM Tris, pH 7.4, 500mM NaCl, 10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
59kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Insect
Lyophilized
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA
HA
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.