missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA Recombinant Influenza A Virus Protein Subtype H4N8 (A/Chicken/Alabama/1/1975), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/chicken/Alabama/1/1975(H4N8) P19695 (Met 1-Ile528) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 40025V08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15286445

  • £211.96 / 50µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Hemagglutinin A (HA), recombinant H4N8 (A/chicken/Alabama/1/1975) HA protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Unknown
PAGE
50μg
C-Terminal
<1 EU/μg
Hemagglutinin A
Assay Standard
Mammalian
Lyophilized
59kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
HA
HA
Cell Structure/Organelle Proteins
His Tag
Videos