Learn More
HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/Brisbane/59/2007), His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of influenza A virus, subtype H1N1 (A/Brisbane/59/2007), HA (ACA28844.1) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 11052V08H250
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Subunits HA1 and HA2 remain uncleaved
- Activity: This recombinant protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5 x 50μg | |
Truncated/Recombinant/Cell Surface Protein | |
HA | |
HA | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
59kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Hemagglutinin | |
His Tag | |
Other Proteins | |
Viral Proteins |
For Research Use Only. Not for use in diagnostic procedures.