Sino Biological™ HA Recombinant Influenza A, Subtype H3N2 (A/Perth/16/2009) Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/Perth/16/2009(H3N2)) (Met 1-Trp 530) (ACS71642.1) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 40043V08H250

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15226495

Quantity Price
1 £ 622.0 / 250µg
EU Stock 1
Add to basket


Description and Specification

Specification

Molecular Weight 59kDa
Common Name Hemagglutinin
Endotoxin Concentration <1 EU/μg
Termination C-Terminal
Format Lyophilized
Quantity 5 x 50μg
Expression System Mammalian
Validated Application Assay Standard
Gene Alias HA
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol HA
Purity ≥85% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Purification Method PAGE
Protein Family Cell Structure/Organelle Proteins
Structural Form Truncated/Recombinant

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.