missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ HA Recombinant Influenza A, Subtype H2N2 (A/Canada/720/2005) Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 516) of the influenza hemagglutinin (A/Canada/720/2005 (H2N2)) (AAY28987.1) (HA1+HA2, uncleaved) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 11688V08H50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298865

  • £211.96 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
59kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
HA
HA
Cell Structure/Organelle Proteins
His Tag
Lyophilized
PAGE
50μg
C-Terminal
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.